3-Phenoxypropionsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Penoxypropionic acid, 3-Phenoxypropanoic acid
M-Clarity™ Qualitätsstufe = MQ100
P16001-5G
SIALP16001-5GEA
62.9
EUR
SIALP16001-5G
3-Phenoxypropionsäure, Sigma-Aldrich®
3-Phenoxypropionsäure
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Siedepunkt: 235 °C (1013 hPa) Schmelzpunkt: 92 °C |
MDL:
MFCD00002769 CAS-Nummer: 7170-38-9 EINECS: 230-518-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...