PD-123319 ditrifluoroacetate, Hyrdrat ≥98% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: PD 123,319 di(trifluoroacetate), 1-[[4-(Dimethylamino)-3-methylphenyl]methyl]-5-(diphenylacetyl)-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid ditrifluoroacetate, S-(+)-1-[(4-(Dimethylamino)-3-methylphenyl)methyl]-5-(diphenylacetyl)-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid di(trifluoroacetate) salt

P186-10MG
SIALP186-10MGEA 372 EUR
SIALP186-10MG
PD-123319 ditrifluoroacetate, Hyrdrat ≥98% (durch HPLC), Sigma-Aldrich®
PD-123319 ditrifluoroacetate
Formel: C₃₁H₃₂N₄O₃·2C₂HF₃O₂·xH₂O
Molecular Weight: 736,66 g/mol (anhy)
MDL: MFCD11046022
CAS-Nummer: 136676-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR