9-Phenylcarbazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Phenylcarbazole

M-Clarity™ Qualitätsstufe = MQ100

P21501-10G
SIALP21501-10GEA 106 EUR
SIALP21501-10G
9-Phenylcarbazol, Sigma-Aldrich®
9-Phenylcarbazole
Formel: C₁₈H₁₃N
Molecular Weight: 243,30 g/mol
MDL: MFCD00004965
CAS-Nummer: 1150-62-5
EINECS: 214-564-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR