1,3-Phenylendiessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2'-(1,3-Phenylene)diacetic acid

P23350-5G
SIALP23350-5GEA 55.2 EUR
SIALP23350-5G
1,3-Phenylendiessigsäure, Sigma-Aldrich®
1,3-Phenylendiessigsäure
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
MDL: MFCD00004341
CAS-Nummer: 19806-17-8
EINECS: 243-332-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR