DL-α-Phenylglycin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dl-A-Aminophenylacetic Acid, H-DL-Phg-OH, DL-2-Phenylglycine

M-Clarity™ Qualitätsstufe = MQ200

P25507-100G
SIALP25507-100GEA 50.8 EUR
SIALP25507-100G
DL-α-Phenylglycin, Sigma-Aldrich®
DL-α-Phenylglycin
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00064402
CAS-Nummer: 2835-06-5
EINECS: 220-608-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR