D(-)-Penicillamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine

MQ100

P4875-1G P4875-25G P4875-5G
SIALP4875-1GEA 60.2 EUR
SIALP4875-1G SIALP4875-25G SIALP4875-5G
D(-)-Penicillamin, Sigma-Aldrich®
D(-)-Penicillamin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149.2 g/mol
MDL: MFCD00064302
CAS-Nummer: 52-67-5
EINECS: 200-148-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR