3,4-(Methylendioxy)benzylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Piperonylamine, Benzo[d][1,3]dioxol-5-ylmethanamine

M-Clarity™ Qualitätsstufe = MQ100

P49503-25G
SIALP49503-25GEA 174 EUR
SIALP49503-25G
3,4-(Methylendioxy)benzylamin, Sigma-Aldrich®
3,4-(Methylendioxy)benzylamin
Formel: C₈H₉NO₂
Molecular Weight: 151,17 g/mol
MDL: MFCD00005840
CAS-Nummer: 2620-50-0
EINECS: 220-056-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR