Propargylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Propinylammoniumchlorid, Propargylammoniumchlorid, Monopropargylamin Hydrochlorid, Prop-2-ynylamin Hydrochlorid, 3-Aminopropin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

P50919-10G
SIALP50919-10GEA 188 EUR
SIALP50919-10G
Propargylamin Hydrochlorid, Sigma-Aldrich®
Propargylamin Hydrochlorid
Formel: C₃H₆ClN
Molecular Weight: 91,54 g/mol
Schmelzpunkt: 178…179 °C
MDL: MFCD00012907
CAS-Nummer: 15430-52-1
EINECS: 239-440-5
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR