Propargylbromid ∼9.2 mol/l in Toluol ≥80%, Sigma-Aldrich®

Lieferant: Merck

P51001-5KG P51001-125G P51001-50G P51001-625G
SIALP51001-5KGEA 4890 EUR
SIALP51001-5KG SIALP51001-125G SIALP51001-50G SIALP51001-625G
Propargylbromid ∼9.2 mol/l in Toluol ≥80%, Sigma-Aldrich®
Propargylbromid
Formel: C₃H₃Br
MDL: MFCD00000241
CAS-Nummer: 106-96-7
EINECS: 203-447-1
UN: 2924
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR