Pluronic® F-68 10% aqueous solution, BioReagent Cell Culture, Filter sterile, Sigma-Aldrich®

Lieferant: Merck

Synonyme: PEG-PPG-PEG, Kolliphor® P 188, Synperonic® F68, Lutrol® F68, Pluronic® F-68

MQ300

P5556-100ML
SIALP5556-100MLEA 52.2 EUR
SIALP5556-100ML
Pluronic® F-68 10% aqueous solution, BioReagent Cell Culture, Filter sterile, Sigma-Aldrich®
Kolliphor® P 188 (Lutrol® F68, Pluronic® F-68, Synperonic® F68)

Poloxamer 188 (P188) is a non-ionic amphiphilic copolymer. It comprises a central chain of hydrophobic polyoxypropylene flanked at ends by hydrophilic polyoxyethylene.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Molecular Weight: 8400
Lagertemperatur: Raumtemperatur
MDL: MFCD00082049
CAS-Nummer: 9003-11-6

Bestellen

Spezifikation Testergebnisse

Appearance (Turbidity) Clear
Appearance (Form) Solution
pH 6.1 - 7.1
Osmolality ≤200 mOs/kg
Sterility by current EP/USP Passes
Endotoxin Level ≤10 EU/ml
Concentration by CDET 9.0 - 11.0 %
Cell Culture Testing - MTT Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR