(R)-(+)-Pulegon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(+)-Pulegone, (R)-2-Isopropylidene-5-methylcyclohexanone, p-Menth-4(8)-en-3-one, (R)-p-Menth-4(8)-en-3-one, (R)-5-Methyl-2-(propan-2-ylidene)cyclohexanone
M-Clarity™ Qualitätsstufe = MQ100
P55708-100ML
SIALP55708-100MLEA
128
EUR
SIALP55708-100ML
(R)-(+)-Pulegon, Sigma-Aldrich®
(R)-(+)-Pulegon
Formel:
C₁₀H₁₆O |
MDL:
MFCD00063000 CAS-Nummer: 89-82-7 EINECS: 201-943-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...