di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP) 40 mg, Kapseln alkalines Phosphatase-Substrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

P5744-25CAP
SIALP5744-25CAPEA 233 EUR
SIALP5744-25CAP
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP) 40 mg, Kapseln alkalines Phosphatase-Substrat, Sigma-Aldrich®
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)

Phosphatase substrate has been used as a substrate in tartrate-resistant acid phosphatase (TRAP) assay and alkaline phosphatase (ALP). Substrate for the determination of acid and alkaline phosphatases.


Chromogenic substrate for use with alkaline and acid phosphatase ELISA procedures. Produces a soluble end product that is yellow in color and can be read spectrophotometrically at 405 nm. The pNPP reaction may be stopped with 3 N NaOH and read at 405 nm.

Formel: C₆H₄NNa₂O₆P·6H₂O
Schmelzpunkt: 300 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00066288
CAS-Nummer: 333338-18-4
EINECS: 224-246-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR