syn-2-Pyridincarbaldehydoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: syn-2-Pyridinealdoxime

M-Clarity™ Qualitätsstufe = MQ100

P58200-25G
SIALP58200-25GEA 227 EUR
SIALP58200-25G
syn-2-Pyridincarbaldehydoxim, Sigma-Aldrich®
syn-2-Pyridincarbaldehydoxim
Formel: C₆H₆N₂O
Molecular Weight: 122,13 g/mol
MDL: MFCD00167166
CAS-Nummer: 1193-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR