Nicotinaldehydoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Pyridinealdoxime, Pyridine-3-carboxaldoxime, Pyridine-3-aldoxime

P58405-25G
SIALP58405-25GEA 48.9 EUR
SIALP58405-25G
Nicotinaldehydoxim, Sigma-Aldrich®
Nicotinaldehydoxim
Formel: C₆H₆N₂O
Molecular Weight: 122,13 g/mol
MDL: MFCD00006409
CAS-Nummer: 1193-92-6
EINECS: 214-782-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR