2,4-Pyridindicarbonsäure Monohydrat ≥98%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Lutidinic acid monohydrate, 2,4-Pyridinedicarboxylic acid monohydrate

M-Clarity™ Qualitätsstufe = MQ100

P63395-5G
SIALP63395-5GEA 82 EUR
SIALP63395-5G
2,4-Pyridindicarbonsäure Monohydrat ≥98%, Sigma-Aldrich®
2,4-Pyridindicarbonsäure Monohydrat
Formel: C₇H₅NO₄·1H₂O
Molecular Weight: 185,14 g/mol
CAS-Nummer: 207671-42-9
EINECS: 207-892-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR