Creatinphosphat Dinatriumsalz Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Phosphocreatine disodium hydrate, Sodium 2-(1-methyl-3-phosphonatoguanidino)acetate hydrate(2:1:x), N-(Imino[phosphonoamino]methyl)-N-methylglycine hydrate, Phosphocreatine disodium salt hydrate

M-Clarity™ Qualitätsstufe = MQ200

P7936-10MG P7936-1G P7936-25G P7936-5G
SIALP7936-10MGEA 41.2 EUR
SIALP7936-10MG SIALP7936-1G SIALP7936-25G SIALP7936-5G
Creatinphosphat Dinatriumsalz Hydrat, Sigma-Aldrich®
Creatinphosphat Dinatriumsalz Hydrat
Formel: C₄H₈N₃Na₂O₅P·H₂O
Molecular Weight: 273.09 g/mol
MDL: MFCD00150191
CAS-Nummer: 19333-65-4
EINECS: 213-074-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR