Gellan Gummi, Sigma-Aldrich®

Lieferant: Merck


MQ200

P8169-100G P8169-1KG P8169-250G P8169-500G P8169-5KG
SIALP8169-100GEA 76 EUR
SIALP8169-100G SIALP8169-1KG SIALP8169-250G SIALP8169-500G SIALP8169-5KG
Gellan Gummi, Sigma-Aldrich®
Gellan Gummi
Lagertemperatur: Raumtemperatur
MDL: MFCD00131909
CAS-Nummer: 71010-52-1
EINECS: 275-117-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR