1,3-Propandisulfonsäure ≥70% (w/v), Sigma-Aldrich®

Lieferant: Merck

P8454-25ML
SIALP8454-25MLEA 234 EUR
SIALP8454-25ML
1,3-Propandisulfonsäure ≥70% (w/v), Sigma-Aldrich®
1,3-Propandisulfonsäure
Formel: C₃H₈O₆S₂
Molecular Weight: 204,21 g/mol
MDL: MFCD00191482
CAS-Nummer: 21668-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR