1,3-Propandisulfonsäure Dinatriumsalz, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium propane-1,3-disulphonate

P8579-25G
SIALP8579-25GEA 102 EUR
SIALP8579-25G
1,3-Propandisulfonsäure Dinatriumsalz, Sigma-Aldrich®
1,3-Propandisulfonsäure Dinatriumsalz
Formel: C₃H₆Na₂O₆S₂
Molecular Weight: 248,19 g/mol
MDL: MFCD00191483
CAS-Nummer: 36589-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR