Polyadenylic acid (poly-A)
Lieferant: Merck
P9403-25MG
P9403-100MG
P9403-250MG
P9403-1G
SIALP9403-25MGEA
119
EUR
SIALP9403-25MG
SIALP9403-100MG
SIALP9403-250MG
SIALP9403-1G
Polyadenylic acid (poly-A)
Vektoren, Plasmide und Bibliotheken
Polyadenylic acid (poly-A) is used to evaluate binding on cationic liposomes doped with non-ionic nucleolipids. Poly-A is used in small molecule mRNA targeted drug development to evaluate the binding of potential therapeutic agents such as the isoquinoline group of alkaloids.