N-(Phosphonomethyl)glycine, BioReagent pflanzliche Zellkulturen getestet

Lieferant: Merck
Gefahr

Synonyme: Glyphosat

M-Clarity™ Qualitätsstufe = MQ200

P9556-5G
SIALP9556-5GEA 680 EUR
SIALP9556-5G
N-(Phosphonomethyl)glycine, BioReagent pflanzliche Zellkulturen getestet
N-(Phosphonomethyl)glycine

Glyphosphate inhibits the shikimate pathway-dependent synthesis of the aromatic amino acids tyrosine, tryptophan and phenylalanine at the growing points of plants.


Glyphosate (N-(phosphonomethyl)glycine) (sold commercially as Roundup) is the most widely used broad-spectrum systemic herbicide in the world. Consequently, the environmental effects of glyphosphate and its mechanism of action are an important area of active research.

Formel: C₃H₈NO₅P
Molecular Weight: 169,07 g/mol
MDL: MFCD00055350
CAS-Nummer: 1071-83-6
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to Off-White
Appearance (Form) Powder
Solubility (Color) (50 mg/mL, 1N NaOH) Colorless to Faint Yellow
Solubility (Turbidity) (50 mg/mL, 1N NaOH) Clear to Slightly Hazy
Infrared spectrum Conforms to Structure
Phosphorus (P) 17.4 - 19.2 %
Plant Cell Culture Test Pas

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR