Procain Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Diethylaminoethyl 4-aminobenzoate hydrochloride
M-Clarity™ Qualitätsstufe = MQ200
P9879-100G
P9879-250G
P9879-50G
SIALP9879-100GEA
65.5
EUR
SIALP9879-100G
SIALP9879-250G
SIALP9879-50G
Procain Hydrochlorid, Sigma-Aldrich®
Procain Hydrochlorid
Formel:
C₁₃H₂₁ClN₂O₂ Molecular Weight: 272.77 g/mol |
MDL:
MFCD00013000 CAS-Nummer: 51-05-8 EINECS: 200-077-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...