IPTG (Isopropyl-β-D-thiogalactopyranosid), Pulver

Lieferant: Merck

Synonyme: IPTG

PHG0010-5G
SIALPHG0010-5GEA 0 EUR
SIALPHG0010-5G
IPTG (Isopropyl-β-D-thiogalactopyranosid), Pulver
IPTG (Isopropyl-β-D-thiogalactopyranosid)
Formel: C₉H₁₈O₅S
Molecular Weight: 238.31 g/mol
Siedepunkt: 402
MDL: MFCD00063273
CAS-Nummer: 367-93-1

Bestellen

Spezifikation Testergebnisse

Quality level 300


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR