Rifampicin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5,6,9,17,19,21-Hexahydroxy-23-methoxy- 2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl- 1-piperazinyl)formimidoyl]-2,7-(epoxypentadeca- [1,11,13]trienimino)-naphtho[2,1-b]furan- 1,11(2H)-dion-21-acetat, Rifampin

M-Clarity™ Qualitätsstufe = MQ200

R7382-1G R7382-5G
SIALR7382-1GEA 208 EUR
SIALR7382-1G SIALR7382-5G
Rifampicin, Sigma-Aldrich®
Rifampicin
Formel: C₄₃H₅₈N₄O₁₂
Schmelzpunkt: 183…188 °C
MDL: MFCD00151389
CAS-Nummer: 13292-46-1
EINECS: 236-312-0
Merck Index: 12,08382

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR