Resorcinmonoacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Hydroxyphenyl acetate, Acetic acid 3-Hydroxyphenyl ester
M-Clarity™ Qualitätsstufe = MQ100
R856-100G
SIALR856-100GEA
59.2
EUR
SIALR856-100G
Resorcinmonoacetat, Sigma-Aldrich®
Resorcinmonoacetat
Formel:
C₈H₈O₃ |
MDL:
MFCD00002266 CAS-Nummer: 102-29-4 EINECS: 203-022-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...