Human IgG ELISA kit
Lieferant: Merck
RAB0001-1KT
SIALRAB0001-1KTEA 608 EUR
SIALRAB0001-1KT
Human IgG ELISA kit
Tests ELISAs
Human IgG ELISA Kit is an in vitro enzyme-linked immunosorbent assay used for the quantitative measurement of a target protein in biological samples, such as serum, plasma, cell culture supernatants, urine, and/or cell and tissue lysates.
Used for the quantitative measurement of a target protein in biological samples, such as serum, plasma, cell culture supernatants, urine, and/or cell and tissue lysates.
Lieferumfang: Kit includes human IgG antibody-coated ELISA plate 96 wells (12 strips×8 wells) , 20X wash buffer concentrated (25 ml), lyophilised human IgG protein standard (2 vials), ELISA 5X assay/sample diluent buffer E, biotinylated human IgG detection antibody (2 vials), HRP-streptavidin (200 µl), ELISA colorimetric TMB reagent-HRP substrate (12 ml of 3, 3', 5, 5'-tetramethylbenzidine in buffer solution) and ELISA stop solution.