D(−)-Salicin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
α-Hydroxy-o-tolyl-β-D-glucopyranosid, (2R3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxan-3,4,5-triol, Weidenbitter, Salicinum, Saligenin-β-D-glucopyranosid, 2-(Hydroxymethyl)phenyl-beta-D-glucopyranosid, Salicosid
M-Clarity™ Qualitätsstufe = MQ200
S0625-100G
S0625-25G
S0625-5G
SIALS0625-100GEA
1060
EUR
SIALS0625-100G
SIALS0625-25G
SIALS0625-5G
D(−)-Salicin, Sigma-Aldrich®
D(−)-Salicin
Formel:
C₁₃H₁₈O₇ Molecular Weight: 286.28 g/mol Schmelzpunkt: 199…201 °C Dichte: 1.42 g/cm³ (20 °C) |
MDL:
MFCD00006590 CAS-Nummer: 138-52-3 EINECS: 205-331-6 Merck Index: 12,08476 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...