Scopoletin (7-Hydroxy-6-methoxy-2H-chromen-2-on), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one

M-Clarity™ Qualitätsstufe = MQ100

S2500-100MG S2500-1G S2500-500MG S2500-50MG
SIALS2500-100MGEA 286 EUR
SIALS2500-100MG SIALS2500-1G SIALS2500-500MG SIALS2500-50MG
Scopoletin (7-Hydroxy-6-methoxy-2H-chromen-2-on), Sigma-Aldrich®
Scopoletin (7-Hydroxy-6-methoxycumarin)
Formel: C₁₀H₈O₄
Molecular Weight: 192.1681 g/mol
MDL: MFCD00006872
CAS-Nummer: 92-61-5
EINECS: 202-171-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR