Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine
M-Clarity™ Qualitätsstufe = MQ200
S4264-1G
S4264-5G
SIALS4264-1GEA
68.3
EUR
SIALS4264-1G
SIALS4264-5G
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin), Sigma-Aldrich®
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin)
Formel:
C₁₀H₂₆N₄ |
MDL:
MFCD00008215 CAS-Nummer: 71-44-3 EINECS: 200-754-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...