Sonnenblumenkernöl, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

S5007-1L S5007-250ML
SIALS5007-1LEA 245 EUR
SIALS5007-1L SIALS5007-250ML
Sonnenblumenkernöl, Sigma-Aldrich®
Sunflower seed oil
MDL: MFCD00132403
CAS-Nummer: 8001-21-6
EINECS: 232-273-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR