Span® 20, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sorbitan monododecanoate, Arlacel 20, Sorbitan monolaurate

M-Clarity™ Qualitätsstufe = MQ200

S6635-1L S6635-250ML
SIALS6635-1LEA 221 EUR
SIALS6635-1L SIALS6635-250ML
Span® 20, Sigma-Aldrich®
Span® 20
Formel: C₁₈H₃₄O₆
Molecular Weight: 346.46 g/mol
Dichte: 1.05 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005365
CAS-Nummer: 1338-39-2
EINECS: 215-663-3
Merck Index: 12,08872

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR