Span® 80, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Arlacel 80, Sorbitan monooleate
M-Clarity™ Qualitätsstufe = MQ200
S6760-1L
SIALS6760-1LEA
208
EUR
SIALS6760-1L
Span® 80, Sigma-Aldrich®
Span® 80
Formel:
C₂₄H₄₄O₆ Molecular Weight: 428,61 g/mol Dichte: 1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00080948 CAS-Nummer: 1338-43-8 EINECS: 215-665-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...