Sigmatrix Urine Diluent
Lieferant: Merck
Sigmatrix urine diluent is a non-biological diluent that mimics human urine.This product can be used as a diluent for sample prep, controls and calibrators and is recommended for LC-MS based applications.
- Used in enzymatic hydrolysis of drug glucuronides and LC-MS analysis of drugs and drug metabolites
- The formulation is dye-free
Buffered solution containing calcium chloride, magnesium chloride, potassium chloride, sodium chloride, sodium phosphate, sodium sulfate, urea, and creatinine with sodium azide as a preservative.
Normal precautions exercised in handling laboratory reagents should be followed. Dispose of waste observing all local, state, provincial, or national regulations.
Biological source: Synthetic
Form: Liquid
Shipped in: Wet ice
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...