Resiquimod, weiß, Pulver

Lieferant: Merck

Synonyme: 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, 1-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-alpha,alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R-848

SML0196-10MG
SIALSML0196-10MGEA 117 EUR
SIALSML0196-10MG
Resiquimod, weiß, Pulver
Resiquimod
Formel: C₁₇H₂₂N₄O₂
Molecular Weight: 314,389 g/mol
MDL: MFCD00937759
CAS-Nummer: 144875-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR