3,3',5,5'-Tetramethylbenzidin (TMB) substrate system, Liquid ELISA, Sigma-Aldrich®

Lieferant: Merck

Synonyme: TMB

T0440-100ML
SIALT0440-100MLEA 209 EUR
SIALT0440-100ML
3,3',5,5'-Tetramethylbenzidin (TMB) substrate system, Liquid ELISA, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidin (TMB)

TMB is a substrate for horseradish peroxidase.


Develops a soluble blue reaction product that may be read at 370 or 655 nm. For endpoint assays the reaction may be stopped with acid, forming a yellow reaction product that may be read at 450 nm. Recommended for ELISA (microwell) procedures, not recommended for membrane applications. 3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA has been used as a substrate for horseradish peroxidase antibody conjugate for detecting hendra virus glycoprotein, tetracosactide, sIgA and neutrophil extracellular traps (NETs).

Formel: C₁₆H₂₀N₂
Molecular Weight: 240.35 g/mol
Siedepunkt: 402
Schmelzpunkt: 166…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007748
CAS-Nummer: 54827-17-7
EINECS: 259-364-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR