1,3,4,6-Tetrachloro-3-6-diphenylglycouril

Lieferant: Merck

Synonyme: Iodo-Gen®, NSC 4462

M-Clarity™ Qualitätsstufe = MQ100

T0656-250MG
SIALT0656-250MGEA 271 EUR
SIALT0656-250MG
1,3,4,6-Tetrachloro-3-6-diphenylglycouril
1,3,4,6-Tetrachloro-3-6-diphenylglycouril

Iodination reagent used in radioiodination acting as an oxidative agent


Reagent involved in comparison studies with chloramine-T
Reagent used for oxidation of urazoles


Bitte beachten Iodo-Gen is a registered trademark of Pierce Biotechnology, Inc.

Formel: C₁₆H₁₀Cl₄N₄O₂
Molecular Weight: 432,088 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00027367
CAS-Nummer: 51592-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR