Tetrahydropyran-2-methanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxymethyltetrahydropyran, 2-(Hydroxymethyl)tetrahydro-2H-pyran

M-Clarity™ Qualitätsstufe = MQ100

T14605-5G
SIALT14605-5GEA 45.2 EUR
SIALT14605-5G
Tetrahydropyran-2-methanol, Sigma-Aldrich®
Tetrahydropyran-2-methanol
Formel: C₆H₁₂O₂
Molecular Weight: 116,16 g/mol
MDL: MFCD00006624
CAS-Nummer: 100-72-1
EINECS: 202-882-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR