Triethanolamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

T1502-100G T1502-1KG T1502-250G T1502-500G
SIALT1502-100GEA 58 EUR
SIALT1502-100G SIALT1502-1KG SIALT1502-250G SIALT1502-500G
Triethanolamin Hydrochlorid, Sigma-Aldrich®
Triethanolamin Hydrochlorid
Formel: C₆H₁₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00012596
CAS-Nummer: 637-39-8
EINECS: 211-284-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR