Tetrahydroxy-1,4-benzoquinone hydrate ≥99%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Tetrahydroxyquinone, 2,3,5,6-Tetrahydroxycyclohexa-2,5-diene-1,4-dione xhydrate, Tetroquinone

M-Clarity™ Qualitätsstufe = MQ100

T17000-5G
SIALT17000-5GEA 268 EUR
SIALT17000-5G
Tetrahydroxy-1,4-benzoquinone hydrate ≥99%, Sigma-Aldrich®
Tetrahydroxy-1,4-benzoquinone Hydrate

Used as a effector of xanthine oxidase.


Synonyms: Tetrahydroxy-1,4-benzoquinone

Formel: C₆H₆O₇
Molecular Weight: 190.11 g/mol
Schmelzpunkt: >300 °C (Lit.)
MDL: MFCD00001597
CAS-Nummer: 123334-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR