Tetramethylensulfoxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrahydrothiophen-1-oxid, Thiacyclopentan-1-oxid

M-Clarity™ Qualitätsstufe = MQ100

T22403-25G
SIALT22403-25GEA 80 EUR
SIALT22403-25G
Tetramethylensulfoxid, Sigma-Aldrich®
Tetramethylensulfoxid
Formel: C₄H₈OS
Molecular Weight: 104,17 g/mol
MDL: MFCD00005477
CAS-Nummer: 1600-44-8
EINECS: 216-493-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR