Natriumtetraphenylborat, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt
M-Clarity™ Qualitätsstufe = MQ200
T25402-100G
T25402-25G
T25402-5G
SIALT25402-100GEA
308
EUR
SIALT25402-100G
SIALT25402-25G
SIALT25402-5G
Natriumtetraphenylborat, Sigma-Aldrich®
Sodium tetraphenylborate
Formel:
(C₆H₅)₄BNa Molecular Weight: 342.22 g/mol Schmelzpunkt: > 400 °C Dichte: 1.15 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011494 CAS-Nummer: 143-66-8 EINECS: 205-605-5 UN: 2811 ADR: 6.1 III Merck Index: 13,08765 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...