Benzo[b]thiophen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Thianaphthene, 1-Benzothiophene

M-Clarity™ Qualitätsstufe = MQ100

T27405-100G T27405-25G
SIALT27405-100GEA 217 EUR
SIALT27405-100G SIALT27405-25G
Benzo[b]thiophen, Sigma-Aldrich®
Benzo[b]thiophen
Formel: C₈H₆S
Molecular Weight: 134.2 g/mol
MDL: MFCD00005864
CAS-Nummer: 95-15-8
EINECS: 202-395-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR