4-(2-Thienyl)buttersäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Thiophen-2-yl)butanoic acid

M-Clarity™ Qualitätsstufe = MQ100

T27804-10G
SIALT27804-10GEA 128 EUR
SIALT27804-10G
4-(2-Thienyl)buttersäure, Sigma-Aldrich®
4-(2-Thienyl)buttersäure
Formel: C₈H₁₀O₂S
Molecular Weight: 170.23 g/mol
MDL: MFCD00005463
CAS-Nummer: 4653-11-6
EINECS: 225-090-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR