2-Thioxo-4-imidazolidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Thioxoimidazolidin-4-one

M-Clarity™ Qualitätsstufe = MQ200

T30406-25G
SIALT30406-25GEA 170 EUR
SIALT30406-25G
2-Thioxo-4-imidazolidinon, Sigma-Aldrich®
2-Thiohydantoin
Formel: C₃H₄N₂OS
Molecular Weight: 116,14 g/mol
MDL: MFCD00005277
CAS-Nummer: 503-87-7
EINECS: 207-977-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR