(3E)-Testosterone-O-(carboxymethyl)oxime : BSA ≥98% (durch TLC), BSA, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

T3392-10MG
SIALT3392-10MGEA 304 EUR
SIALT3392-10MG
(3E)-Testosterone-O-(carboxymethyl)oxime : BSA ≥98% (durch TLC), BSA, Sigma-Aldrich®
(3E)-Testosterone-O-(carboxymethyl)oxime : BSA

Drug control: Regulated under CDSA - not available from Sigma-Aldrich Canada


Storage temp. : 2-8°C

Formel: C₂₁H₃₁NO₄
Molecular Weight: 361,482 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00132414
CAS-Nummer: 10190-93-9

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Lyophilised powder
Appearance (Colour) White
Solubility (Solvent) Phosphate buffer
Solubility (pH) 7,50 - 7,50
Solubility (Conc.) 1,90 - 2,10 mg/ml
Solubility (Turbidity) Clear to slightly hazy
Solubility (Colour) Colourless
Purity (TLC) ≥98,00%
Conjugation (Ratio Steroid/BSA) 20,00 - 30,00 mole/m
Storage temperature 2 - 8 °C
Extent of labeling 20 - 30 mol steroid per mol BSA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR