p-Toluylsäurehydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: p-Toluoylhydrazine, 4-Methylbenzohydrazide, p-Toluic acid hydrazide, p-Toluic Hydrazide

M-Clarity™ Qualitätsstufe = MQ100

T37001-25G
SIALT37001-25GEA 108 EUR
SIALT37001-25G
p-Toluylsäurehydrazid, Sigma-Aldrich®
p-Toluylsäurehydrazid
Formel: C₈H₁₀N₂O
Molecular Weight: 150.18 g/mol
MDL: MFCD00007607
CAS-Nummer: 3619-22-5
EINECS: 222-815-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR