2,4,5,6-Tetraaminopyrimidinsulfat ≥97%, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

T3807-25G
SIALT3807-25GEA 40.4 EUR
SIALT3807-25G
2,4,5,6-Tetraaminopyrimidinsulfat ≥97%, Sigma-Aldrich®
2,4,5,6-Tetraaminopyrimidinsulfat
Formel: C₄H₈N₆·H₂SO₄
Molecular Weight: 238,23 g/mol
MDL: MFCD00012786
CAS-Nummer: 5392-28-9
EINECS: 226-393-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR