m-Tolylhydraziniumchlorid ≥99%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3-Methylphenylhydrazin Hydrochlorid, 1-(3-Methylphenyl)hydrazin Hydrochlorid, 3-Tolylhydrazin Hydrochlorid, 3-Methylphenylhydraziniumchlorid, 1-(3-Tolyl)hydrazin Hydrochlorid, 3-Tolylhydrazinhydrochlorid, 3-Methylphenylhydrazinhydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

T40401-10G
SIALT40401-10GEA 151 EUR
SIALT40401-10G
m-Tolylhydraziniumchlorid ≥99%, Sigma-Aldrich®
m-Tolylhydraziniumchlorid
Formel: C₇H₁₀N₂·HCl
Molecular Weight: 158,63 g/mol
MDL: MFCD00012932
CAS-Nummer: 637-04-7
EINECS: 211-276-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR