Tetradecyltrimethylammoniumbromid, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

T4762-100G T4762-1KG T4762-250G T4762-500G T4762-5G
SIALT4762-100GEA 123 EUR
SIALT4762-100G SIALT4762-1KG SIALT4762-250G SIALT4762-500G SIALT4762-5G
Tetradecyltrimethylammoniumbromid, Sigma-Aldrich®
Tetradecyltrimethylammoniumbromid
Formel: C₁₇H₃₈BrN
Schmelzpunkt: 245…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011770
CAS-Nummer: 1119-97-7
EINECS: 214-291-9
UN: 3261
ADR: 8,II
Merck Index: 12,06419

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR