Triethylendimin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trientine HCl, N1,N1'-(Ethane-1,2-diyl)bis(ethane-1,2-diamine) dihydrochloride

M-Clarity™ Qualitätsstufe = MQ200

T5033-25G
SIALT5033-25GEA 318 EUR
SIALT5033-25G
Triethylendimin Dihydrochlorid, Sigma-Aldrich®
Triethylendimin Dihydrochlorid
Formel: C₆H₂₀Cl₂N₄
Molecular Weight: 219,16 g/mol
MDL: MFCD00012889
CAS-Nummer: 38260-01-4
EINECS: 253-854-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR